Learn More
Ace Glass, Inc. 14.5mm x 51cm chromatography column
SDP
Catalog No. 50978588
Encompass_Preferred
14.5mm x 51cm chromatography column with 2mm bore 1:5 taper solid PTFE stopcock plug at the bottom.
Catalog No. 50-978-588 Supplier Ace Glass, Inc. Supplier No. 588820
14.5mm x 51cm chromatography column with 2mm bore 1:5 taper solid PTFE stopcock plug at the bottom.