Learn More
Medchemexpress LLC 14-azido-3,6,9,12-tetraoxatetradecan-1-ol | 86770-68-5 | 97.0% | 263.29 g/mol | C10H21N3O5 | 1 G
SDP
Catalog No. 5000203145
Encompass_Preferred
Azido-PEG5-alcohol is a polyethylene glycol (PEG) linker with a terminal azide and a terminal alcohol, used for bioconjugation and linker installation. The azide group enables copper-catalyzed and strain-promoted azide-alkyne cycloaddition reactions, while the PEG5 spacer improves solubility and flexibility for conjugation workflows.
  • Terminal azide enables CuAAC and SPAAC click reactions.
  • PEG5 spacer increases solubility and reduces steric hindrance.
  • Suitable for use as a non-cleavable linker in conjugation chemistry.
  • Colorless to light yellow liquid with high purity.
  • Stable under recommended storage conditions for long-term use.
Catalog No. 50-002-03145 Supplier Medchemexpress LLC Supplier No. HY1302111G
Azido-PEG5-alcohol is a polyethylene glycol (PEG) linker with a terminal azide and a terminal alcohol, used for bioconjugation and linker installation. The azide group enables copper-catalyzed and strain-promoted azide-alkyne cycloaddition reactions, while the PEG5 spacer improves solubility and flexibility for conjugation workflows.
  • Terminal azide enables CuAAC and SPAAC click reactions.
  • PEG5 spacer increases solubility and reduces steric hindrance.
  • Suitable for use as a non-cleavable linker in conjugation chemistry.
  • Colorless to light yellow liquid with high purity.
  • Stable under recommended storage conditions for long-term use.