Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 14-Azido-3,6,9,12-Tetraoxatetradecanoic Acid Solution

Catalog No. 744751500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-475-1500MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 74-475-1500MG Supplier Sigma Aldrich Supplier No. 744751500MG
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

14-Azido-3,6,9,12-Tetraoxatetradecanoic Acid Solution

Specifications

CAS 1334177-78-4
Molecular Formula C26H24F10O9
Linear Formula C26H24F10O9
MDL Number MFCD18916983
Quantity 100 mg
Synonym 2-{2-{2-[2-(2-Azidoethoxy)ethoxy]ethoxy}ethoxy}acetic acid solution; N33-TEG-COOH
Molecular Weight (g/mol) 670.45 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.