Learn More
Medchemexpress LLC HY-N6839 5mg Medchemexpress, 1,4-b-D-Xylopentaose CAS:49694-20-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN68395MG
Encompass_Preferred
Medchemexpress, HY-N6839 5mg 1,4-b-D-Xylopentaose CAS:49694-20-4 1,4-b-D-Xylopentaose (Xylopentaose) consists of five b-1,4 xylose sugars. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2968