Learn More
Sigma Aldrich Fine Chemicals Biosciences 14 Butanedithiol 97 FG3KG
Catalog No. 502907693
Encompass
1 4-Butanedithiol an alkanedithiol is a food odorant. Its odor threshold is similar to its other homologs (propane-1 3-dithiol and pentane-1 5-dithiol).
Catalog No. 50-290-7693 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W5208023KG
1 4-Butanedithiol an alkanedithiol is a food odorant. Its odor threshold is similar to its other homologs (propane-1 3-dithiol and pentane-1 5-dithiol).