Learn More
Medchemexpress LLC HY-N1490 1mg Medchemexpress, 14-Deoxy-11,12-didehydroandrographolide CAS:42895-58-9 Purity:>98%
SDP
Catalog No. 501933843
Encompass_Preferred
Medchemexpress, HY-N1490 1mg 14-Deoxy-11,12-didehydroandrographolide CAS:42895-58-9 14-Deoxy-11,12-didehydroandrographolide is an analogue of Andrographolide that can be isolated from A. paniculata. 14-Deoxy-11,12-didehydroandrographolide inhibits NF-κB activation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3843 Supplier Medchemexpress LLC Supplier No. HYN14901MG
Medchemexpress, HY-N1490 1mg 14-Deoxy-11,12-didehydroandrographolide CAS:42895-58-9 14-Deoxy-11,12-didehydroandrographolide is an analogue of Andrographolide that can be isolated from A. paniculata. 14-Deoxy-11,12-didehydroandrographolide inhibits NF-κB activation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.