Learn More
Medchemexpress LLC 14-Deoxy-11,12-didehydroandrographolide | 42895-58-9 | 332.43 | 1 ML
SDP
Catalog No. 5000415647
Encompass_Preferred
14-Deoxy-11,12-didehydroandrographolide is an analogue of Andrographolide that inhibits NF-κB activation. This naturally occurring, noncytotoxic compound effectively reduces Ovalbumin (OVA)-induced inflammatory cell recruitment, cytokine production, and serum IgE synthesis in a mouse asthma model. It also addresses pulmonary eosinophilia, mucus hypersecretion, mast cell degranulation, and airway hyper-responsiveness.
  • Reduces inflammation.
  • Decreases allergic responses.
  • Improves respiratory function in asthma models.
  • Exhibits noncytotoxic properties.
Catalog No. 50-004-15647 Supplier Medchemexpress LLC Supplier No. HYN149010MM/1ML
14-Deoxy-11,12-didehydroandrographolide is an analogue of Andrographolide that inhibits NF-κB activation. This naturally occurring, noncytotoxic compound effectively reduces Ovalbumin (OVA)-induced inflammatory cell recruitment, cytokine production, and serum IgE synthesis in a mouse asthma model. It also addresses pulmonary eosinophilia, mucus hypersecretion, mast cell degranulation, and airway hyper-responsiveness.
  • Reduces inflammation.
  • Decreases allergic responses.
  • Improves respiratory function in asthma models.
  • Exhibits noncytotoxic properties.