Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 14-Deoxy-11,12-didehydroandrographolide | 42895-58-9 | 332.43 | 25 MG
SDP

Catalog No. 5000389251
Encompass_Preferred

14-Deoxy-11,12-didehydroandrographolide is an analogue of Andrographolide and inhibits NF-κB activation. It is for research use only.

  • Analogue of Andrographolide
  • Inhibits NF-κB activation
  • Naturally occurring noncytotoxic analogue
  • Effectively reduces ovalbumin (OVA)-induced inflammatory cell recruitment into bronchoalveolar lavage (BAL) fluid, IL-4, IL-5, IL-13, and eotaxin production, serum IgE synthesis, pulmonary eosinophilia, mucus hypersecretion, mast cell degranulation, and airway hyper-responsiveness (AHR) in a mouse asthma model, possibly via inhibition of NF-κB activity
  • Dramatically reduces resistance (Rl) and restores Cdyn in OVA-challenged mice in response to methacholine
  • Purity: 99.93%
  • Appearance: Solid
  • Color: White to light yellow
  • Available with data sheet, COA, SDS, and handling instructions

Catalog No. 50-003-89251 Supplier Medchemexpress LLC Supplier No. 25MGHYN1490
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

14-Deoxy-11,12-didehydroandrographolide is an analogue of Andrographolide and inhibits NF-κB activation. It is for research use only.

  • Analogue of Andrographolide
  • Inhibits NF-κB activation
  • Naturally occurring noncytotoxic analogue
  • Effectively reduces ovalbumin (OVA)-induced inflammatory cell recruitment into bronchoalveolar lavage (BAL) fluid, IL-4, IL-5, IL-13, and eotaxin production, serum IgE synthesis, pulmonary eosinophilia, mucus hypersecretion, mast cell degranulation, and airway hyper-responsiveness (AHR) in a mouse asthma model, possibly via inhibition of NF-κB activity
  • Dramatically reduces resistance (Rl) and restores Cdyn in OVA-challenged mice in response to methacholine
  • Purity: 99.93%
  • Appearance: Solid
  • Color: White to light yellow
  • Available with data sheet, COA, SDS, and handling instructions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.