Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 14-Deoxy-11,12-didehydroandrographolide | 42895-58-9 | MFCD07778081 | 332.43 | 100 MG
SDP

Catalog No. 5000414990
Encompass_Preferred

14-Deoxy-11,12-didehydroandrographolide is an analogue of Andrographolide. It inhibits NF-κB activation.

  • An analogue of Andrographolide.
  • Inhibits NF-κB activation.
  • Reduces ovalbumin (OVA)-induced inflammatory cell recruitment into bronchoalveolar lavage (BAL) fluid, IL-4, IL-5, IL-13, eotaxin production, serum IgE synthesis, pulmonary eosinophilia, mucus hypersecretion, mast cell degranulation, and airway hyper-responsiveness (AHR) in a mouse asthma model.
  • At 1 mg/kg, it dramatically reduces resistance (Rl) and restores Cdyn in OVA-challenged mice in response to methacholine.
  • Appearance: Solid.
  • Color: White to light yellow.
  • Purity: 99.93%.

Catalog No. 50-004-14990 Supplier Medchemexpress LLC Supplier No. HYN1490100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

14-Deoxy-11,12-didehydroandrographolide is an analogue of Andrographolide. It inhibits NF-κB activation.

  • An analogue of Andrographolide.
  • Inhibits NF-κB activation.
  • Reduces ovalbumin (OVA)-induced inflammatory cell recruitment into bronchoalveolar lavage (BAL) fluid, IL-4, IL-5, IL-13, eotaxin production, serum IgE synthesis, pulmonary eosinophilia, mucus hypersecretion, mast cell degranulation, and airway hyper-responsiveness (AHR) in a mouse asthma model.
  • At 1 mg/kg, it dramatically reduces resistance (Rl) and restores Cdyn in OVA-challenged mice in response to methacholine.
  • Appearance: Solid.
  • Color: White to light yellow.
  • Purity: 99.93%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.