Learn More
Medchemexpress LLC 14-deoxy-11,12-didehydroandrographolide | 42895-58-9 | MFCD07778081 | 99.6% | 332.4 g/mol | C20H28O4 | 10 MG
SDP
Catalog No. 5000295954
Encompass_Preferred
14-Deoxy-11,12-didehydroandrographolide is a diterpene lactone analogue of andrographolide used as a research reagent. It has been reported to inhibit NF-κB activation and is applied in studies of inflammation and cancer signaling pathways.
  • Diterpene lactone with molecular formula C20H28O4.
  • Reported inhibitor of NF-κB activation in cellular assays.
  • High purity suitable for analytical and biological studies.
  • Available in small research pack sizes (for example, 10 mg) for screening and assay development.
Catalog No. 50-002-95954 Supplier Medchemexpress LLC Supplier No. HYN149010MG
14-Deoxy-11,12-didehydroandrographolide is a diterpene lactone analogue of andrographolide used as a research reagent. It has been reported to inhibit NF-κB activation and is applied in studies of inflammation and cancer signaling pathways.
  • Diterpene lactone with molecular formula C20H28O4.
  • Reported inhibitor of NF-κB activation in cellular assays.
  • High purity suitable for analytical and biological studies.
  • Available in small research pack sizes (for example, 10 mg) for screening and assay development.