Learn More
Sigma Aldrich Fine Chemicals Biosciences 14 Dimethoxybenzene 99 fG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences W23860010KGK
Encompass
1 4-Dimethoxybenzene can be used as a flavoring agent in the food industry. It s one of the main volatile constituents identified inTuber brumale Vietnamese green tea and lotus tea.
Catalog No. 50-290-6470