Learn More
Sigma Aldrich Fine Chemicals Biosciences 14-Dioxane-d8 99 atom D1G
Catalog No. 502891485
Encompass
1,4-Dioxane-d8 is a deuterated derivative of 1,4-dioxane. It has an isotopic purity of 99atom%D. It is a standard purity solvent suitable for routine NMR analyses (conducted at ambient temperatures where quality is less critical). It is widely employed as a deuterated standard. It participates as a surrogate standard during the activated carbon SPE (solid-phase extraction) and GC-MS-SIM (gas chromatography-mass spectrometry (GC-MS) in selected ion monitoring (SIM) mode) quantification of 1,4-dioxane in drinking water.
Catalog No. 50-289-1485 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1864061G
1,4-Dioxane-d8 is a deuterated derivative of 1,4-dioxane. It has an isotopic purity of 99atom%D. It is a standard purity solvent suitable for routine NMR analyses (conducted at ambient temperatures where quality is less critical). It is widely employed as a deuterated standard. It participates as a surrogate standard during the activated carbon SPE (solid-phase extraction) and GC-MS-SIM (gas chromatography-mass spectrometry (GC-MS) in selected ion monitoring (SIM) mode) quantification of 1,4-dioxane in drinking water.