Learn More
Sigma Aldrich Fine Chemicals Biosciences 14-Dioxane suitable for HP1L
Catalog No. 502897476
Encompass
1 4-Dioxane is widely used as a solvent for various organic compounds in industry. It is a non-biodegradable contaminant found in industrial effluents. Its oxidative degradation by using oxygen and ozone has been useful in converting it into biodegradable products which can be easily removed by conventional methods.
Catalog No. 50-289-7476 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 348571L
1 4-Dioxane is widely used as a solvent for various organic compounds in industry. It is a non-biodegradable contaminant found in industrial effluents. Its oxidative degradation by using oxygen and ozone has been useful in converting it into biodegradable products which can be easily removed by conventional methods.