Learn More
Medchemexpress LLC HY-137239 5mg Medchemexpress, 1,4-DPCA ethyl ester CAS:86443-19-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1372395MG
Encompass_Preferred
Medchemexpress, HY-137239 5mg 1,4-DPCA ethyl ester CAS:86443-19-8 1,4-DPCA ethyl ester is the ethyl ester of 1,4-DPCA and can inhibit factor inhibiting HIF (FIH). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1480