Learn More
Electron Microscopy Sciences 14% Ethyl Alcohol, Denatured, 37.5% Formalin 37-39%, 6.25% Glacial Acetic Acid, and D.I. Water
SDP
Catalog No. NC3867479
Encompass
This is rapid fixative. Results with good nuclear detail and minimal formalin pigment. Hatmann's (Davidson's) Fixative is useful for preparing tumors, bone marrow, gynecologic specimen, fatty breast, medical biopsies. Fixation of small specimen is rapid. Exposure to the fixative should be limited to 24 hours (tissue may then be transferred to neutral buffered formalin or 70% alcohol for storage). Based on the volume of 1125 ml solution, there is 2 part Formalin (35-39%), 3 part Ethanol, 1 part Glacial Acetic acid, and 3 part of tap water.
Catalog No. NC3867479 Supplier Electron Microscopy Sciences Supplier No. 64133501G
This is rapid fixative. Results with good nuclear detail and minimal formalin pigment. Hatmann's (Davidson's) Fixative is useful for preparing tumors, bone marrow, gynecologic specimen, fatty breast, medical biopsies. Fixation of small specimen is rapid. Exposure to the fixative should be limited to 24 hours (tissue may then be transferred to neutral buffered formalin or 70% alcohol for storage). Based on the volume of 1125 ml solution, there is 2 part Formalin (35-39%), 3 part Ethanol, 1 part Glacial Acetic acid, and 3 part of tap water.