Learn More
Brady™ SPC™ Absorbents 14 Gallon Universal Spill Kit Refill
Catalog No. 17000908 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English) Sorbing:
12 gal.
Catalog No. Capacity (English) Sorbing
17-000-908 12 gal.
1 options
Catalog No. 17-000-908 Supplier Brady™ SPC™ Absorbents Supplier No. SKA14R
  • $142.70
    $111.65 / Each of 1
    Save $31.05
    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Refill your spill kit's absorbents, accessories, PPE and instructional guidelines so you are prepared for when a spill occurs.
SPC™ Spill Kits from Brady™ combine absorbents, accessories, PPE and instructional guidelines so you are prepared for when a spill occurs. Improve worker productivity and efficiency by placing spill kits around your facility close to areas prone to spills.
  • Refill kit only (container sold separately)
  • Ideal for fast response to critical accidents
Specifications
Capacity (English) Reservoir 14 gal.
Capacity (English) Sorbing 12 gal.
Capacity (Metric) Reservoir 53 L
Capacity (Metric) Sorbing 45.4 L
Material Polypropylene
Type Spill Kit
For Use With (Application) Absorbents and Spill Control
Includes 20 Universal Pads (15 x 19 in.), 2 Socks (3 in. x 12 ft.), 2 Disposal Bags, 1 Pair of Goggles, 1 Pair of Nitrile Gloves