Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vileda Professional™ 14-inch Original Roll-O-Matic™ Refills

Catalog No. 19200720 Shop All Vileda Professional Products
Change view
Click to view available options
Autoclavable:
Autoclavable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Autoclavable
19-200-720 Autoclavable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-200-720 Supplier Vileda Professional™ Supplier No. 142058

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Refills are ideal for use in ISO 6, LAR, Production, and Lab environments

  • Fits all 14 in. Roll-O-Matic™ Handles
  • Ideal for the Pull And Lift technique most commonly used in controlled environments
  • Fully autoclavable
  • Bulk packed
  • Galvanized channel

Specifications

Product Type Sponge Mop Refill
Color Blue
Sterility Non-sterile
Autoclavable Autoclavable
Size 14 in.
Quantity 60/Cs.
For Use With (Application) Floors, walls and ceilings in Iso 6, LAR, Production and Lab Environments
Material Foam
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.