Learn More
Cayman Chemical 14-methyl PalmItIc AcId 10mg
SDP
Catalog No. 502932506
Encompass_Preferred
A methylated fatty acid; has been found in bacteria, bovine milk fat, Aegean jellyfish, and one-humped camel meat and fat; found in human breast milk and levels are decreased in mature breast milk compared to colostrum; produced in Florida manatee liver in response to brevetoxin exposure
Catalog No. 50-293-2506 Supplier Cayman Chemical Supplier No. 2482010 MG
A methylated fatty acid; has been found in bacteria, bovine milk fat, Aegean jellyfish, and one-humped camel meat and fat; found in human breast milk and levels are decreased in mature breast milk compared to colostrum; produced in Florida manatee liver in response to brevetoxin exposure