Learn More
Cayman Chemical 14-methyl Palmitic Acid, Purity: ≥98%, 20 mg
SDP
Catalog No. 502932505
Encompass_Preferred
A methylated fatty acid; has been found in bacteria, bovine milk fat, Aegean jellyfish, and one-humped camel meat and fat; found in human breast milk and levels are decreased in mature breast milk compared to colostrum; produced in Florida manatee liver in response to brevetoxin exposure
Catalog No. 50-293-2505 Supplier Cayman Chemical Supplier No. 2482020 MG
A methylated fatty acid; has been found in bacteria, bovine milk fat, Aegean jellyfish, and one-humped camel meat and fat; found in human breast milk and levels are decreased in mature breast milk compared to colostrum; produced in Florida manatee liver in response to brevetoxin exposure