Learn More
Cayman Chemical 14-Anhydrodigitoxigenin, Purity: ≥98%, CAS: 4321-20-4, 50 mg
SDP
Catalog No. 502990843
Encompass_Preferred
A cardenolide; reduces the activity of guinea pig heart Na+/K+-ATPase by 15% at 10 µM
Catalog No. 50-299-0843 Supplier Cayman Chemical Supplier No. 3043950 MG
A cardenolide; reduces the activity of guinea pig heart Na+/K+-ATPase by 15% at 10 µM