Learn More
Express Diagnostics 14 PANEL CLIA TEST CUP 25/BX
SDP
Supplier:  Express Diagnostics 613075
Encompass_Preferred
14 panel Clia Waived test cup. The Clia Waived test that detects 14 panels for the possible use of:AMP 500BAR 300BUP 10BZO 300COC 150MDMA 500MET 500MTD 300OPI 300OXY 100PCP 25PPX 300TCA 1000THC 50Adulterants – CR, NI, pH, Bl, S.G.CLIA Waived Test 14 panel is for healthcare professionals including professionals at point of care sites. For in-vitro diagnostic use only. The innovative drug test cup design eliminates urine handling and donor tampering, providing a unique and efficient approach for on-site drug testing.
Catalog No. NC1480652
 Restricted Item