Learn More
Stanco Manufacturing 14″ PBI/Kevlar Blend Glove
Catalog No. NC1881895
Encompass_Preferred
Made from 22 oz. ceramic and Kevlar Blend material. The best material for high temperature hand protection. All styles are fully wool lined, extra large cut, and sewn with 100% Kevlar thread. 1,350°F Max Temperature Range.
Catalog No. NC1881895 Supplier Stanco Manufacturing Supplier No. KP214DWL
Made from 22 oz. ceramic and Kevlar Blend material. The best material for high temperature hand protection. All styles are fully wool lined, extra large cut, and sewn with 100% Kevlar thread. 1,350°F Max Temperature Range.