Learn More
Cayman Chemical 14-Pentadecenoic Acid, Purity: ≥98%, CAS: 17351-34-7, 100 mg
SDP
Catalog No. 502981573
Encompass_Preferred
A 15-carbon, long-chain fatty acid that contains an alkene functional group on the terminal carbon of its aliphatic tail
Catalog No. 50-298-1573 Supplier Cayman Chemical Supplier No. 19724100 MG
A 15-carbon, long-chain fatty acid that contains an alkene functional group on the terminal carbon of its aliphatic tail