Learn More
Cayman Chemical 14-PentadecenoIc AcId 10mg
SDP
Catalog No. 502984488
Encompass_Preferred
A 15-carbon, long-chain fatty acid that contains an alkene functional group on the terminal carbon of its aliphatic tail
Catalog No. 50-298-4488 Supplier Cayman Chemical Supplier No. 1972410 MG
A 15-carbon, long-chain fatty acid that contains an alkene functional group on the terminal carbon of its aliphatic tail