Learn More
Cayman Chemical 14-TrIamcInolon acetonIde 10mg
SDP
Catalog No. 502894233
Encompass_Preferred
A potential impurity found in commercial triamcinolone acetonide preparations
Catalog No. 50-289-4233 Supplier Cayman Chemical Supplier No. 900008710 MG
A potential impurity found in commercial triamcinolone acetonide preparations