Learn More
Chemglass Life Sciences Flask Support Ring, Cork, 140 mm OD x 85 mm ID for Round-Bottom Flasks
SDP
Catalog No. NC3037301
Encompass_Preferred
Cork flask support ring designed to stabilize and cushion round-bottom flasks during routine laboratory operations. The dense cork construction provides a non-abrasive seating surface that helps protect glassware and maintain steady support on benchtops or lab stands.
  • Outer diameter: 140 mm
  • Inner diameter: 85 mm
  • Material: cork
  • Provides cushioned, non-abrasive support for round-bottom flasks
  • Sold individually
Catalog No. NC3037301 Supplier Chemglass Life Sciences Supplier No. CG3060140
Cork flask support ring designed to stabilize and cushion round-bottom flasks during routine laboratory operations. The dense cork construction provides a non-abrasive seating surface that helps protect glassware and maintain steady support on benchtops or lab stands.
  • Outer diameter: 140 mm
  • Inner diameter: 85 mm
  • Material: cork
  • Provides cushioned, non-abrasive support for round-bottom flasks
  • Sold individually