Learn More
Selleck Chemical LLC 1400W 2HCl 10mg 214358-33-5 N-(3-(Aminomethyl)benzyl)acetamidine
SDP
Catalog No. 501365963
Encompass_Preferred
1400W (N-(3-(Aminomethyl)benzyl)acetamidine) is a slow, tight binding, and highly selective inhibitor of inducible nitric-oxide synthase(iNOS). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5963 Supplier Selleck Chemical LLC Supplier No. S833710MG
1400W (N-(3-(Aminomethyl)benzyl)acetamidine) is a slow, tight binding, and highly selective inhibitor of inducible nitric-oxide synthase(iNOS). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.