Learn More
Selleck Chemical LLC 1400W 2HCl
SDP
Catalog No. 5000000640
Encompass_Preferred
1400W (N-(3-(Aminomethyl)benzyl)acetamidine) is a slow tight binding and highly selective inhibitor of inducible nitric-oxide synthase(iNOS)
Catalog No. 50-000-00640 Supplier Selleck Chemical LLC Supplier No. S8337200MG
1400W (N-(3-(Aminomethyl)benzyl)acetamidine) is a slow tight binding and highly selective inhibitor of inducible nitric-oxide synthase(iNOS)