Learn More
AdipoGen 1400W . dihydrochloride (25 mg)
SDP
Supplier:  AdipoGen AGCR10018M025
Encompass_Preferred
Chemical. CAS: 214358-33-5. Formula: C10H15N3 . 2HCl. MW: 177.3 . 72.9. A slow, tight binding and highly selective inhibitor of iNOS (inducible nitric oxide synthase/NOS II) (Kd < 7 nM). Weak and reversible inhibition of nNOS (neuronal nitric oxide synthase/NOS I) (Ki < 2) and eNOS (endothelial nitric oxide synthase/NOS III) (Ki < 50 µM). Inhibits tumor growth. Increases vasoconstriction to noradrenaline. Improves contractile function. Improves stroke outcome and decreases glutamate release. Anti-inflammatory. Review.
Catalog No. 50-205-3766