Learn More
Sino Biological Recombinant Human MOB4A/MOB1B Protein (GST Tag)
Supplier:  Sino Biological 14016H09E100
Encompass_Preferred
A DNA sequence encoding the human MOBKL1A (Q7L9L4) (Met1-Arg216) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-8988