Learn More
Sino Biological Recombinant Human RPRD1B Protein (His Tag)
Supplier:  Sino Biological 14027H07H100
Encompass_Preferred
A DNA sequence encoding the human RPRD1B (AAH33629.1) (Met1-Asp326) was expressed with an N-terminal polyhistidine tag.
Catalog No. 50-161-8992