Learn More
Sino Biological Recombinant Human PEX11A Protein (mFc Tag)
Supplier:  Sino Biological 14029H04H100
Encompass_Preferred
A DNA sequence encoding the human PEX11A (NP_003838.1) (Arg106-Pro219) was expressed with the Fc region of mouse IgG1 at the N-terminus.
Catalog No. 50-161-8994