Learn More
Sino Biological Recombinant Human TM4SF2/TSPAN7 Protein (His Tag)
Supplier:  Sino Biological 14031H08H100
Encompass_Preferred
A DNA sequence encoding the human TSPAN7 (AAH18036.1) (Arg113-Met213) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8998