Learn More
Sino Biological Recombinant Human DAP10 Protein (mFc Tag)
Supplier:  Sino Biological 14063H05H100
Encompass_Preferred
A DNA sequence encoding the human HCST (Q9UBK5-1) (Met-Pro48) was fused with Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-9000