Learn More
eMolecules​ 141-43-5 | 2-Aminoethanol, redistilled | Oakwood Chemicals | MFCD00008183 | 61.084 | C2H7NO | 99.000 | NCCO | 500g | 486563294
Catalog No. 5000490603 Shop All eMolecules Inc. Products
Encompass_Preferred
2-Aminoethanol, redistilled | Oakwood Chemicals | 141-43-5 | MFCD00008183 | 61.084 | C2H7NO | 99.000 | NCCO | 500g | 486563294
Catalog No. 50-004-90603 Supplier eMolecules​ Supplier No. 486563294
2-Aminoethanol, redistilled | Oakwood Chemicals | 141-43-5 | MFCD00008183 | 61.084 | C2H7NO | 99.000 | NCCO | 500g | 486563294