Learn More
Sino Biological Recombinant Human NDRG1 Protein (His Tag)
Supplier:  Sino Biological 14119H08E100
Encompass_Preferred
A DNA sequence encoding the human NDRG1 (Q92597) (Met1-Cys394) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9010