Learn More
Sino Biological Recombinant Human HBP1 Protein (GST Tag)
Supplier:  Sino Biological 14127H09E100
Encompass_Preferred
A DNA sequence encoding the mature form of human HBP1 (O60381-1) (Pro208-Phe345) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-9012