Learn More
Sino Biological Recombinant Human LSM1 Protein (His Tag)
Supplier:  Sino Biological 14142H07E100
Encompass_Preferred
A DNA sequence encoding the mature form of human LSM1 (O15116) (Met1-Tyr133) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-9020