Learn More
Sino Biological Recombinant Human PDRG1 Protein (His Tag)
Supplier:  Sino Biological 14143H07E100
Encompass_Preferred
A DNA sequence encoding the human PDRG1 (Q9NUG6) (Met1-Gly133) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9022