Learn More
Sino Biological Recombinant Human B3GALT5 Protein (His Tag)
Supplier:  Sino Biological 14155H07B100
Encompass_Preferred
A DNA sequence encoding the human B3GALT5 (NP_006048.1) (Asn29-Val310) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9028