Learn More
Sino Biological Recombinant Human PPIL1 Protein (His Tag)
Supplier:  Sino Biological 14163H08E100
Encompass_Preferred
A DNA sequence encoding the human PPIL1 (Q9Y3C6) (Met1-Gly166) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9030