Learn More
Sino Biological Recombinant Human Aspartate Aminotransferase Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 14196H07E100
Encompass_Preferred
A DNA sequence encoding the mature form of human GOT1 (P17174) (Met1-Gln413) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9038