Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GVS AcetatePlus™ 142 mm Cellulose Acetate Disc

Catalog No. 09924041 Shop All GVS Products
Change view
Click to view available options
Pore Size:
0.22 μm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size
09-924-041 0.22 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-924-041 Supplier GVS AcetatePlus™ Supplier No. 1215074
Add to Cart
Add to Cart

Cellulose acetate is a supported, hydrophilic membrane that is naturally low binding. Available from 25 mm to 293 mm. It is ideal for filtration applications where maximal recovery of protein is critical.

Properties

  • Membranes are composed of pure cellulose acetate that is internally supported by an inert polyester web. This web gives each membrane exceptional strength to prevent cracking, tearing, breaking and distortion when handled or creased.
  • The resulting membrane has dimensional stability that can withstand autoclaving or steam sterilizing leaving the membrane unaffected in temperatures up to 274°F (135°C).
  • The exceptional dimensional strength and low binding characteristics of the cellulose acetate filtration membranes provides higher throughputs than competitive offerings and reduces the amount of filter changes needed during proteinaceous solution filtering.

Characteristics

  • Superior strength can withstand aggressive handling or be used with automated equipment without breaking or tearing
  • Low extractables ensures tests will be clean with consistent results
  • Hydrophilic wets out rapidly.
  • Lot-to-lot consistency quality checks ensure consistent flow and diffusion rates for dependable results every time.
  • Nonlysing of cells prevents contamination of critical solutions.
  • Pore size range: 0.1 μm to 20 μm

Applications

  • Biological fluid sterilization
  • Cold sterilization
  • Protein and enzyme filtration
  • Tissue culture media sterilization

Specifications

Autoclavable Autoclavable
Disposable Limited-use
Filter Type Membrane Filter
Format Disc
Material Cellulose Acetate
Pore Size 0.22 μm
Sterility Non-sterile
Type Disc Membrane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.