Learn More
Sino Biological Recombinant Human Iba1 Protein (His Tag)
Supplier:  Sino Biological 14227H07E100
Encompass_Preferred
A DNA sequence encoding the mature form of human AIF1 (P55008-1) (Met1-Pro147) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-9046