Learn More
Sino Biological Recombinant Human ARHI Protein (mFc Tag)
Supplier:  Sino Biological 14239H04H100
Encompass_Preferred
A DNA sequence encoding the human DIRAS3 (O95661) (Met1-Lys225) was expressed with the Fc region of mouse IgG1 at the N-terminus.
Catalog No. 50-161-9052