Learn More
Sino Biological Recombinant Human Syntaxin 8 Protein (His Tag)
Supplier:  Sino Biological 14243H07H100
Encompass_Preferred
A DNA sequence encoding the human STX8 (Q9UNK0) (Met1-Gly215) was expressed with an N-terminal polyhistidine tag.
Catalog No. 50-161-9054