Learn More
Sino Biological Recombinant Human CD81 Protein (ECD, His Tag)
Supplier:  Sino Biological 14244H07H100
Encompass_Preferred
A DNA sequence encoding the human CD81 (NP_004347.1) (Phe113-Lys201) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9058