Learn More
Sino Biological Recombinant Human ESD Protein (His Tag)
Supplier:  Sino Biological 14252H07E100
Encompass_Preferred
A DNA sequence encoding the mature form of human ESD (P10768) (Met1-Ala282) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-9062