Learn More
Sino Biological Recombinant Human UMP-CMP kinase/CMPK1 Protein (His Tag)
Supplier:  Sino Biological 14258H07B50
Encompass_Preferred
A DNA sequence encoding the human CMPK1 (P30085) (Met1-Gly196) was fused with a polyhistide tag at the N-terminus.
Catalog No. 50-161-9066