Learn More
Sino Biological Recombinant Human CKAP1/TBCB Protein (His Tag)
Supplier:  Sino Biological 14260H07E100
Encompass_Preferred
A DNA sequence encoding the mature form of human TBCB (Q99426) (Met1-Ile244) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-9067